Skip to Content
Merck
Fmoc-Cys(Acm)-OH Novabiochem®

Fmoc-Cys(Acm)-OH

Fmoc-Cys(Acm)-OH Novabiochem®
Synonym(s):
Fmoc-Cys(Acm)-OH
Empirical Formula (Hill Notation):
C21H22N2O5S
CAS Number:
Molecular Weight:
414.47

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Functional Group
Application
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing