Fexofenadine impurity A,
Synonym(s):
4-[4-[4-(Hydroxydiphenylmethyl)-1-piperidinyl]-1-oxobutyl]-α,α-dimethylphenylacetic acid
Empirical Formula (Hill Notation):
C32H37NO4
CAS Number:
Molecular Weight:
499.64
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing