![Fenthion-sulfone certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/192/258/eb54df62-a776-4927-bd45-81399d577d78/640/eb54df62-a776-4927-bd45-81399d577d78.png)
Fenthion-sulfone
![Fenthion-sulfone certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/192/258/eb54df62-a776-4927-bd45-81399d577d78/640/eb54df62-a776-4927-bd45-81399d577d78.png)
Empirical Formula (Hill Notation):
C10H15O5PS2
CAS Number:
Molecular Weight:
310.33
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2