Ethyl 2-methylpentanoate
Synonym(s):
Manzanate
Linear Formula:
CH3CH2CH2CH(CH3)CO2C2H5
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing