![Enoxolone for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/130/041/3085d104-ee47-482c-af15-dcfee7fed4b2/640/3085d104-ee47-482c-af15-dcfee7fed4b2.png)
Enoxolone for peak identification
![Enoxolone for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/130/041/3085d104-ee47-482c-af15-dcfee7fed4b2/640/3085d104-ee47-482c-af15-dcfee7fed4b2.png)
Synonym(s):
18β-Glycyrrhetinic acid, 3β-Hydroxy-11-oxo-18β,20β-olean-12-en-29-oic acid, Enoxolone, Glycyrrhetin, Subglycyrrhelinic acid, Uralenic acid
Empirical Formula (Hill Notation):
C30H46O4
CAS Number:
Molecular Weight:
470.68
Products
Formula Weight
Manufacturer
Melting Point (°C)
Application
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing