Skip to Content
Merck
Dofetilide Related Compound A certified reference material, pharmaceutical secondary standard

Dofetilide Related Compound A

Dofetilide Related Compound A certified reference material, pharmaceutical secondary standard
Synonym(s):
N-{4-{2-{2-[4-(Methanesulfonamido)phenoxy]ethyl}amino}phenyl}methanesulfonamide, N-Desmethyldofetilide
Empirical Formula (Hill Notation):
C18H25N3O5S2
CAS Number:
Molecular Weight:
427.54

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade