![DMT-2′O-Methyl-rC(ac) Phosphoramidite configured for PerkinElmer, configured for Polygen](/deepweb/assets/sigmaaldrich/product/structures/646/889/7761e925-9a43-41c3-bde0-07655be98192/640/7761e925-9a43-41c3-bde0-07655be98192.png)
DMT-2′O-Methyl-rC(ac) Phosphoramidite
![DMT-2′O-Methyl-rC(ac) Phosphoramidite configured for PerkinElmer, configured for Polygen](/deepweb/assets/sigmaaldrich/product/structures/646/889/7761e925-9a43-41c3-bde0-07655be98192/640/7761e925-9a43-41c3-bde0-07655be98192.png)
Synonym(s):
N-acetyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-methyl-cytidine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]
Empirical Formula (Hill Notation):
C42H52N5O9P
CAS Number:
Molecular Weight:
801.86
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing