
(±)-6-Chloro-PB hydrobromide

Synonym(s):
(±)-6-Chloro-7,8-dihydroxy-1-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepine hydrobromide, (±)-SKF-81297 hydrobromide
Empirical Formula (Hill Notation):
C16H16ClNO2 · HBr
CAS Number:
Molecular Weight:
370.67
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing