Skip to Content
Merck
Diniconazole certified reference material, TraceCERT®, E-Isomer, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Diniconazole

Diniconazole certified reference material, TraceCERT®, E-Isomer, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Empirical Formula (Hill Notation):
C15H17Cl2N3O
CAS Number:
Molecular Weight:
326.22

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Quality Segment
Reference Material Type
Special Grade