![Diflufenzopyr sodium salt PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/455/660/744240e2-fa01-44f7-84f3-6edcc5127655/640/744240e2-fa01-44f7-84f3-6edcc5127655.png)
Diflufenzopyr sodium salt
![Diflufenzopyr sodium salt PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/455/660/744240e2-fa01-44f7-84f3-6edcc5127655/640/744240e2-fa01-44f7-84f3-6edcc5127655.png)
Synonym(s):
2-{1-(3,5-Difluorophenylaminocarbonylhydrazono)ethyl}-3-pyridinecarboxylic acid monosodium salt
Empirical Formula (Hill Notation):
C15H11F2N4NaO3
CAS Number:
Molecular Weight:
356.26
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing