Skip to Content
Merck
Dibenzo[b,e]thiepin-11(6H)-one British Pharmacopoeia (BP) Reference Standard

Dibenzo[b,e]thiepin-11(6H)-one

Dibenzo[b,e]thiepin-11(6H)-one British Pharmacopoeia (BP) Reference Standard
Synonym(s):
6,11-Dihydrodibenzo[b,e]thiepin-11-one
Empirical Formula (Hill Notation):
C14H10OS
CAS Number:
Molecular Weight:
226.29

Products

Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing