
Di-2-thienyl ketone

Empirical Formula (Hill Notation):
C9H6OS2
CAS Number:
Molecular Weight:
194.27
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing