![D-Homoserine](/deepweb/assets/sigmaaldrich/product/structures/106/895/6b0b5aa7-48f4-40e7-bd5b-2b554d3f8c4a/640/6b0b5aa7-48f4-40e7-bd5b-2b554d3f8c4a.png)
D-Homoserine
![D-Homoserine](/deepweb/assets/sigmaaldrich/product/structures/106/895/6b0b5aa7-48f4-40e7-bd5b-2b554d3f8c4a/640/6b0b5aa7-48f4-40e7-bd5b-2b554d3f8c4a.png)
Synonym(s):
(R)-(+)-2-Amino-4-hydroxybutyric acid
Linear Formula:
HOCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
119.12
Products
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Optical Isomer
Amino Acid
Showing 1-1 of 1
Compare
Product Number
Description
Pricing