Skip to Content
Merck
Dexamethasone acetate for peak identification CRS European Pharmacopoeia (EP) Reference Standard

Dexamethasone acetate for peak identification CRS

Dexamethasone acetate for peak identification CRS European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Dexamethasone 21-acetate, 9α-Fluoro-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione-21-acetate, 9α-Fluoro-16α-methylprednisolone-21-acetate, Prednisolone F acetate
Empirical Formula (Hill Notation):
C24H31FO6
CAS Number:
Molecular Weight:
434.50

Products

Feature
Formula Weight
Manufacturer
Application
Special Grade
Showing 1-1 of 1