![Desoximetasone United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/768/849/869f53c7-ff35-46b5-8398-5d47b7bc0377/640/869f53c7-ff35-46b5-8398-5d47b7bc0377.png)
Desoximetasone
![Desoximetasone United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/768/849/869f53c7-ff35-46b5-8398-5d47b7bc0377/640/869f53c7-ff35-46b5-8398-5d47b7bc0377.png)
Synonym(s):
9-Fluoro-11β,21-dihydroxy-16α-methylpregna-1,4-diene-3,20-dione
Empirical Formula (Hill Notation):
C22H29FO4
CAS Number:
Molecular Weight:
376.46
Products
Biological Source
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing