![Creatine phosphate crystalline, >97%](/deepweb/assets/sigmaaldrich/product/structures/265/272/698f2b50-c584-4bff-a25a-5fc6cbdbc075/640/698f2b50-c584-4bff-a25a-5fc6cbdbc075.png)
Creatine phosphate
![Creatine phosphate crystalline, >97%](/deepweb/assets/sigmaaldrich/product/structures/265/272/698f2b50-c584-4bff-a25a-5fc6cbdbc075/640/698f2b50-c584-4bff-a25a-5fc6cbdbc075.png)
Synonym(s):
Sodium creatine phosphate dibasic tetrahydrate, Creatine phosphate disodium salt tetrahydrate
Linear Formula:
Na2O3PNHC(=NH)N(CH3)CH2COOH · 4H2O
CAS Number:
Molecular Weight:
327.14
Products
Formula Weight
Manufacturer
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing