Clomiphene Related Compound A
Synonym(s):
(E/Z)-2-[4-(1,2-Diphenylethenyl)phenoxy]-N,N-diethylethanamine hydrochloride
Empirical Formula (Hill Notation):
C26H29NO · HCl
CAS Number:
Molecular Weight:
407.98
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2