Clarithromycin Identity
Synonym(s):
Clarithromycin
Empirical Formula (Hill Notation):
C38H69NO13
CAS Number:
Molecular Weight:
747.95
Products
Product Category
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing