Skip to Content
Merck
cis-8,11,14-Eicosatrienoic acid methyl ester ≥99%

cis-8,11,14-Eicosatrienoic acid methyl ester

cis-8,11,14-Eicosatrienoic acid methyl ester ≥99%
Synonym(s):
Dihomo-γ-linolenic acid methyl ester, Fame 20:3n-6, Methyl DGLA
Empirical Formula (Hill Notation):
C21H36O2
CAS Number:
Molecular Weight:
320.51

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing