Skip to Content
Merck
Chlortetracycline hydrochloride British Pharmacopoeia (BP) Reference Standard

Chlortetracycline hydrochloride

Chlortetracycline hydrochloride British Pharmacopoeia (BP) Reference Standard
Synonym(s):
7-Chlorotetracycline hydrochloride, Chlortetracycline hydrochloride
Empirical Formula (Hill Notation):
C22H23ClN2O8 · HCl
CAS Number:
Molecular Weight:
515.34
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
Package Size
API Family
Reference Material Type
Special Grade
Technique