Skip to Content
Merck
Carbetamide PESTANAL®, analytical standard

Carbetamide

Carbetamide PESTANAL®, analytical standard
Synonym(s):
(R)-(−)-1-(Ethylcarbamoyl)ethyl N-phenylcarbamate
Empirical Formula (Hill Notation):
C12H16N2O3
CAS Number:
Molecular Weight:
236.27
EC Number:

Products

Product Category
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing