![Candesartan Cilexetil Related Compound G Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/566/571/3052b5e6-6a79-4dc0-b7ef-e7fcce62056a/640/3052b5e6-6a79-4dc0-b7ef-e7fcce62056a.jpg)
Candesartan cilexetil Related Compound G
![Candesartan Cilexetil Related Compound G Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/566/571/3052b5e6-6a79-4dc0-b7ef-e7fcce62056a/640/3052b5e6-6a79-4dc0-b7ef-e7fcce62056a.jpg)
Synonym(s):
Candesartan, 2-Ethoxy 1-{[2′-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl}benzimidazole-7-carboxylic acid
Empirical Formula (Hill Notation):
C24H20N6O3
CAS Number:
Molecular Weight:
440.45
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2