Skip to Content
Merck
Cadmium chloride technical grade

Cadmium chloride

Cadmium chloride technical grade
Synonym(s):
Cadmium dichloride, Dichlorocadmium
Empirical Formula (Hill Notation):
CdCl2
Molecular Weight:
183.32
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Reagent Type
Core
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing