Skip to Content
Merck
Butylhydroxyanisole European Pharmacopoeia (EP) Reference Standard

Butylhydroxyanisole

Butylhydroxyanisole European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Butylated hydroxyanisole, 2(3)-t-Butyl-4-hydroxyanisole, 2(3)-t-Butylhydroquinone monomethyl ether, BHA
Linear Formula:
(CH3)3CC6H3(OCH3)OH
CAS Number:
Molecular Weight:
180.24
EC Number:

Products

Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing