
Bulleyaconitine A

Synonym(s):
((1-α,6-α,14-α,16-β)-8-(acetyloxy)-20-ethyl-13-hydroxy-1,6,16-trimethoxy-4-(methoxymethyl)aconitan-14-yl)(4-methoxyphenyl)-methanone
Empirical Formula (Hill Notation):
C35H49NO9
CAS Number:
Molecular Weight:
627.76
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing