![Borane-ammonia complex technical grade, 90%](/deepweb/assets/sigmaaldrich/product/structures/128/533/7c1cdcf7-2944-4e3f-b971-dfe340daf022/640/7c1cdcf7-2944-4e3f-b971-dfe340daf022.png)
Borane-ammonia complex
![Borane-ammonia complex technical grade, 90%](/deepweb/assets/sigmaaldrich/product/structures/128/533/7c1cdcf7-2944-4e3f-b971-dfe340daf022/640/7c1cdcf7-2944-4e3f-b971-dfe340daf022.png)
Synonym(s):
Amminetrihydroboron, Borazane
Linear Formula:
H3N · BH3
CAS Number:
Molecular Weight:
30.87
Products
Brand
Sustainability
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing