Skip to Content
Merck
Boc-Phe-OMe 98%

Boc-Phe-OMe

Boc-Phe-OMe 98%
Synonym(s):
N-(tert-Butoxycarbonyl)-L-phenylalanine methyl ester, Boc-L-phenylalanine methyl ester
Linear Formula:
C6H5CH2CH[NHCO2C(CH3)3]CO2CH3
CAS Number:
Molecular Weight:
279.33

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing