![Boc-β-phenyl-Phe-OH ≥98.0% (TLC)](/deepweb/assets/sigmaaldrich/product/structures/159/717/b6640e12-3633-4626-bc5b-ac28a24e8bf1/640/b6640e12-3633-4626-bc5b-ac28a24e8bf1.png)
Boc-β-phenyl-Phe-OH
![Boc-β-phenyl-Phe-OH ≥98.0% (TLC)](/deepweb/assets/sigmaaldrich/product/structures/159/717/b6640e12-3633-4626-bc5b-ac28a24e8bf1/640/b6640e12-3633-4626-bc5b-ac28a24e8bf1.png)
Synonym(s):
N-(tert-Butoxycarbonyl)-β-phenyl-L-phenylalanine, Boc-β,β-diphenyl-Ala-OH, Boc-3,3-diphenyl-L-alanine
Linear Formula:
(C6H5)2CHCH[NHCO2C(CH3)3]CO2H
CAS Number:
Molecular Weight:
341.40
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing