![Boc-Ala-OH-1-13C 99 atom % 13C](/deepweb/assets/sigmaaldrich/product/structures/398/305/8b352927-2a33-48ac-ae15-5429d8dcd648/640/8b352927-2a33-48ac-ae15-5429d8dcd648.png)
Boc-Ala-OH-1-13C
![Boc-Ala-OH-1-13C 99 atom % 13C](/deepweb/assets/sigmaaldrich/product/structures/398/305/8b352927-2a33-48ac-ae15-5429d8dcd648/640/8b352927-2a33-48ac-ae15-5429d8dcd648.png)
Synonym(s):
N-(tert-Butoxycarbonyl)-L-alanine-1-13C, L-Alanine-1-13C, N-t-Boc derivative
Linear Formula:
CH3CH(NH-Boc)13CO2H
CAS Number:
Molecular Weight:
190.20
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Functional Group
Isotope
Application
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing