Skip to Content
Merck
β-Nicotinamide adenine dinucleotide 2′-phosphate reduced tetrasodium salt hydrate ampule, ≥90% (HPLC)

β-Nicotinamide adenine dinucleotide 2′-phosphate reduced tetrasodium salt hydrate

β-Nicotinamide adenine dinucleotide 2′-phosphate reduced tetrasodium salt hydrate ampule, ≥90% (HPLC)
Synonym(s):
β-NADPH, 2′-NADPH hydrate, Coenzyme II reduced tetrasodium salt, Dihydronicotinamide adenine dinucleotide phosphate tetrasodium salt, NADPH Na4, TPNH2 Na4, Triphosphopyridine nucleotide reduced tetrasodium salt
Empirical Formula (Hill Notation):
C21H26N7Na4O17P3 · xH2O
CAS Number:
Molecular Weight:
833.35 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity