Biperiden hydrochloride
Synonym(s):
1-(5-Bicyclo[2.2.1]hept-2-enyl)-1-phenyl-3-(1-piperidinyl)propan-1-ol hydrochloride, Akineton, Akinophyl
Empirical Formula (Hill Notation):
C21H30ClNO
CAS Number:
Molecular Weight:
347.92
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing