![β-Estradiol-6-one 6-(O-carboxymethyloxime)](/deepweb/assets/sigmaaldrich/product/structures/170/990/4cb312eb-e090-4c41-9069-b7324b592093/640/4cb312eb-e090-4c41-9069-b7324b592093.png)
β-Estradiol-6-one 6-(O-carboxymethyloxime)
![β-Estradiol-6-one 6-(O-carboxymethyloxime)](/deepweb/assets/sigmaaldrich/product/structures/170/990/4cb312eb-e090-4c41-9069-b7324b592093/640/4cb312eb-e090-4c41-9069-b7324b592093.png)
Synonym(s):
β-Estradiol-6-(O-carboxymethyl)oxime, 1,3,5(10)-Estratriene-3,17β-diol-6-one6-(O-carboxymethyloxime), 3,17β-Dihydroxy-1,3,5(10)-estratriene 6-(O-carboxymethyl)oxime, 6-Ketoestradiol 6-(O-carboxymethyloxime)
Empirical Formula (Hill Notation):
C20H25NO5
CAS Number:
Molecular Weight:
359.42
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing