Skip to Content
Merck
Benomyl PESTANAL®, analytical standard

Benomyl

Benomyl PESTANAL®, analytical standard
Synonym(s):
Methyl 1-(butylcarbamoyl)-2-benzimidazolecarbamate, Benomyl, NSC 263489
Empirical Formula (Hill Notation):
C14H18N4O3
CAS Number:
Molecular Weight:
290.32
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing