Skip to Content
Merck
α-Terpinene analytical standard

α-Terpinene

α-Terpinene analytical standard
Synonym(s):
1-Isopropyl-4-methyl-1,3-cyclohexadiene, p-Mentha-1,3-diene
Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing