Anti-NUP88 antibody produced in rabbit
Synonym(s):
Anti-88 kDa nuclear pore complex protein, Anti-Nuclear pore complex protein Nup88, Anti-Nucleoporin Nup88
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1