Skip to Content
Merck
Aminohippuric Acid pharmaceutical secondary standard, certified reference material

Aminohippuric acid

Aminohippuric Acid pharmaceutical secondary standard, certified reference material
Synonym(s):
p-Aminohippuric acid, 4-Aminohippuric acid, N-(4-Aminobenzoyl)glycine
Linear Formula:
NH2C6H4C(O)NHCH2COOH
CAS Number:
Molecular Weight:
194.19

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade