![Ametoctradin hydrochloride Metabolite M650F06 certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/554/469/35904d04-316b-4c31-a637-af4a205efd4c/640/35904d04-316b-4c31-a637-af4a205efd4c.png)
Ametoctradin hydrochloride Metabolite M650F06
![Ametoctradin hydrochloride Metabolite M650F06 certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/554/469/35904d04-316b-4c31-a637-af4a205efd4c/640/35904d04-316b-4c31-a637-af4a205efd4c.png)
Synonym(s):
6-(7-Amino-5-ethyl[1,2,4]triazolo[1,5-a]pyrimidin-6-yl)hexanoic acid hydrochloride
Empirical Formula (Hill Notation):
C13H19N5O2· HCl
CAS Number:
Molecular Weight:
313.78
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing