Skip to Content
Merck
Amantadine Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material

Amantadine Related Compound A

Amantadine Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
1-Chloroadamantane
Empirical Formula (Hill Notation):
C10H15Cl
CAS Number:
Molecular Weight:
170.68

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade