![Aciclovir for impurity C identification CRS, European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/817/179/cdc344db-5da0-49c9-a29b-b025ce3c722e/640/cdc344db-5da0-49c9-a29b-b025ce3c722e.png)
Aciclovir for impurity C identification
![Aciclovir for impurity C identification CRS, European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/817/179/cdc344db-5da0-49c9-a29b-b025ce3c722e/640/cdc344db-5da0-49c9-a29b-b025ce3c722e.png)
Synonym(s):
Acycloguanosine, 9-[(2-Hydroxyethoxy)methyl]guanine, Acyclovir
Empirical Formula (Hill Notation):
C8H11N5O3
CAS Number:
Molecular Weight:
225.20
Products
Formula Weight
Manufacturer
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing