Skip to Content
Merck
Acetylcholine chloride European Pharmacopoeia (EP) Reference Standard

Acetylcholine chloride

Acetylcholine chloride European Pharmacopoeia (EP) Reference Standard
Synonym(s):
ACh, Acetylcholine chloride
Linear Formula:
(CH3)3N+CH2CH2OCOCH3Cl-
CAS Number:
Molecular Weight:
181.66
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Technique