Skip to Content
Merck
Acetyl-6-formylpterin ≥95% (HPLC)

Acetyl-6-formylpterin

Acetyl-6-formylpterin ≥95% (HPLC)
Synonym(s):
2-Acetamido-6-formylpteridin-4-one, N-(6-formyl-4-hydroxy-2-pteridinyl)acetamide, N-(6-formyl-4-oxo-1H-pteridin-2-yl)acetamide
Empirical Formula (Hill Notation):
C9H7N5O3
CAS Number:
Molecular Weight:
233.18

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing