Abacavir Related Compound B
Synonym(s):
[4-(2,5-Diamino-6-chloropyrimidin-4-ylamino)cyclopent-2-enyl]methanol
Empirical Formula (Hill Notation):
C10H14ClN5O
Molecular Weight:
255.70
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2