Skip to Content
Merck
vibrant-m

Abacavir impurity standard

vibrant-m
Synonym(s):
Abacavir sulfate
Empirical Formula (Hill Notation):
C28H36N12O2 · H2SO4
CAS Number:
Molecular Weight:
670.74

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing