8-Hydroxy-3,4-Dihydroquinolin-2(1H)-One
Synonym(s):
8-Hydroxy-3,4-dihydroquinolin-2(1H)-one
Empirical Formula (Hill Notation):
C9H9NO2
CAS Number:
Molecular Weight:
163.17
Products
Brand
Formula Weight
Manufacturer
Product Line
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade