Skip to Content
Merck
8-(4-Chlorophenylthio)adenosine 3′,5′-cyclic monophosphate sodium salt ≥97.0% (HPLC), powder

8-(4-Chlorophenylthio)adenosine 3′,5′-cyclic monophosphate sodium salt

8-(4-Chlorophenylthio)adenosine 3′,5′-cyclic monophosphate sodium salt ≥97.0% (HPLC), powder
Synonym(s):
pCPT-cAMP
Empirical Formula (Hill Notation):
C16H14ClN5NaO6PS
CAS Number:
Molecular Weight:
493.79
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Showing 1-1 of 1