Skip to Content
Merck
5-Chloro-N-Et-4-hydroxy-1-methyl-2-oxo-N-Ph-1,2-dihydroquinoline-3-carboxamide

5-Chloro-N-Et-4-hydroxy-1-methyl-2-oxo-N-Ph-1,2-dihydroquinoline-3-carboxamide

5-Chloro-N-Et-4-hydroxy-1-methyl-2-oxo-N-Ph-1,2-dihydroquinoline-3-carboxamide
Empirical Formula (Hill Notation):
C19H17ClN2O3
CAS Number:
Molecular Weight:
356.80

Products

Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1