5-Carboxy-tetramethylrhodamine
Synonym(s):
5-Carboxy-tetramethylrhodamine, 5-TAMRA
Empirical Formula (Hill Notation):
C25H22N2O5
CAS Number:
Molecular Weight:
430.45
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Application
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing