5-Bromovaleric acid
Synonym(s):
5-Bromovaleric acid
Linear Formula:
Br(CH2)4COOH
CAS Number:
Molecular Weight:
181.03
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing