Skip to Content
Merck
4-Methyl-2-thiazolecarboxaldehyde 97%

4-Methyl-2-thiazolecarboxaldehyde

4-Methyl-2-thiazolecarboxaldehyde 97%
Empirical Formula (Hill Notation):
C5H5NOS
CAS Number:
Molecular Weight:
127.16

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing