Skip to Content
Merck
4-Hexylresorcinol analytical standard

4-Hexylresorcinol

4-Hexylresorcinol analytical standard
Synonym(s):
4-Hexyl-1,3-dihydroxybenzene, 4-Hexylresorcinol
Linear Formula:
CH3(CH2)5C6H3-1,3-(OH)2
CAS Number:
Molecular Weight:
194.27
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing